1-Bromoadamantano ≥99.0% (por GC), purified by sublimation

Proveedor: TCI

B5879-1G B5879-5G
TCIAB5879-1GEA 61 EUR
TCIAB5879-1G TCIAB5879-5G
1-Bromoadamantano ≥99.0% (por GC), purified by sublimation
1-Bromoadamantano
Formula: C₁₀H₁₅Br
Número MDL: MFCD00074721
Núm. CAS: 768-90-1
EINECS: 212-200-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 118.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR