3-Butyn-1-amine hydrochloride ≥95.0% (by titrimetric analysis)

Proveedor: TCI

B5925-1G B5925-200MG
TCIAB5925-1GEA 113 EUR
TCIAB5925-1G TCIAB5925-200MG
3-Butyn-1-amine hydrochloride ≥95.0% (by titrimetric analysis)
3-Butyn-1-amine hydrochloride
Formula: C₄H₇N·HCl
Número MDL: MFCD06658391
Núm. CAS: 88211-50-1

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 220.0 to 224.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR