tert-Butyl Hydrogen Dodecanedioate ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B5926-1G
B5926-5G
TCIAB5926-1GEA
247
EUR
TCIAB5926-1G
TCIAB5926-5G
tert-Butyl Hydrogen Dodecanedioate ≥98.0% (by GC, titration analysis)
tert-Butyl Hydrogen Dodecanedioate
Formula:
C₁₆H₃₀O₄ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD28098251 Núm. CAS: 234081-98-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...