18-(tert-Butoxy)-18-oxooctadecanoic acid ≥95.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
tert-Butyl Hydrogen Octadecanedioate, Mono-tert-butyl Octadecanedioate, tBu ester-C16-acid, Octadecanedioic acid Mono-tert-butyl ester
B5928-1G
B5928-250MG
TCIAB5928-1GEA
236
EUR
TCIAB5928-1G
TCIAB5928-250MG
18-(tert-Butoxy)-18-oxooctadecanoic acid ≥95.0% (by GC, titration analysis)
18-(tert-Butoxy)-18-oxooctadecanoic acid
Formula:
C₂₂H₄₂O₄ |
Número MDL:
MFCD09991735 Núm. CAS: 843666-40-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...