1-Benzyl-3,3-difluoro-4,4-piperidinediol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
1-Benzyl-3,3-difluoropiperidine-4,4-diol
B5930-1G
B5930-200MG
TCIAB5930-1GEA
89
EUR
TCIAB5930-1G
TCIAB5930-200MG
1-Benzyl-3,3-difluoro-4,4-piperidinediol ≥98.0% (by GC, titration analysis)
1-Benzyl-3,3-difluoro-4,4-piperidinediol
Formula:
C₁₂H₁₅F₂NO₂ MW: 243,26 g/mol |
Número MDL:
MFCD20926176 Núm. CAS: 1067914-81-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...