Benzyl N,N,N',N'-Tetraisopropylphosphorodiamidite ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
1-(Benzyloxy)-N,N,N',N'-tetraisopropylphosphinediamine
B5997-1G
B5997-5G
TCIAB5997-1GEA
103
EUR
TCIAB5997-1G
TCIAB5997-5G
Benzyl N,N,N',N'-Tetraisopropylphosphorodiamidite ≥98.0% (by GC, titration analysis)
Benzyl N,N,N',N'-Tetraisopropylphosphorodiamidite
Benzyl N,N,N',N'-tetraisopropylphosphorodiamidite [B5997] is used in the synthesis of phosphodiesters and phosphorylation of inositols and nucleosides.
B5997 promptly reacts with alcohols in the presence of a base like 1H-tetrazole to afford a phosphoramidite. It can further react with another alcohol resulting in a phosphite triester with different alcohol species. Successive oxidation of the phosphorus atom and deprotection gives a phosphodiester.
Formula:
C₁₉H₃₅N₂OP MW: 338.48 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD32062870 Núm. CAS: 108549-21-9 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...