SB-297006 ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: N-Benzoyl-4-nitro-L-phenylalanine Ethyl ester, SB297006, (S)-Ethyl 2-benzamido-3-(4-nitrophenyl)propanoate

B6053-5MG B6053-25MG
TCIAB6053-5MGEA 71 EUR
TCIAB6053-5MG TCIAB6053-25MG
SB-297006 ≥98.0% (by HPLC)
SB-297006
Formula: C₁₈H₁₈N₂O₅
MW: 342,35 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD19690954
Núm. CAS: 58816-69-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 119.0 to 123.0 deg-C
Specific rotation [a]20/D -82.0 to -90.0 deg (C=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR