SB 225002 ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: SB225002, 1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea

B6055-10MG B6055-50MG
TCIAB6055-10MGEA 82 EUR
TCIAB6055-10MG TCIAB6055-50MG
SB 225002 ≥98.0% (by HPLC)
SB 225002
Formula: C₁₃H₁₀BrN₃O₄
MW: 352,10 g/mol
Número MDL: MFCD00954637
Núm. CAS: 182498-32-4

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.30 to 12.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR