2-(2-Bromophenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: 2-(2-Bromophenyl)-1H-1,3-benzodiazole, 2-(2-Bromophenyl)-1H-benzo[d]imidazole

B6092-1G B6092-5G
TCIAB6092-1GEA 31 EUR
TCIAB6092-1G TCIAB6092-5G
2-(2-Bromophenyl)-1H-benzimidazole ≥98.0% (by GC, titration analysis)
2-(2-Bromophenyl)-1H-benzimidazole
Formula: C₁₃H₉BrN₂
Número MDL: MFCD00449969
Núm. CAS: 13275-42-8

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 239.0 to 243.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR