(1R)-Camphor oxime ≥98.0% (por GC)

Proveedor: TCI

C0013-1G C0013-25G C0013-5G
TCIAC0013-1GEA 25 EUR
TCIAC0013-1G TCIAC0013-25G TCIAC0013-5G
(1R)-Camphor oxime ≥98.0% (por GC)
(1R)-Camphor oxime
Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Número MDL: MFCD00046298
Núm. CAS: 2792-42-9
EINECS: 220-525-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4d8cf8a81fca57091b0a4636c85e1629f749f80&vid=b4d8cf8a81fca57091b0a4636c85e1629f749f80&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 120.0 deg-C
Specific rotation [a]20/D -40.0 to -42.0 deg (C=10,EtOH)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR