Hexanamide ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Capronamide
C0024-25G
C0024-5G
TCIAC0024-25GEA
100
EUR
TCIAC0024-25G
TCIAC0024-5G
Hexanamide ≥98.0% (by total nitrogen basis)
Hexanamide
Formula:
C₆H₁₃NO MW: 115,18 g/mol |
Número MDL:
MFCD00008046 Núm. CAS: 628-02-4 EINECS: 211-024-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 98.0 to 101.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...