N-Chloroacetyl-L-valine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
C0107-1G
C0107-100MG
TCIAC0107-1GEA
49
EUR
TCIAC0107-1G
TCIAC0107-100MG
N-Chloroacetyl-L-valine ≥98.0% (by titrimetric analysis)
N-Chloroacetyl-L-valine
Formula:
C₇H₁₂ClNO₃ MW: 193,63 g/mol |
Número MDL:
MFCD00045273 Núm. CAS: 2279-16-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 113.0 to 116.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...