Cholesterol Palmitate ≥95.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Palmitic acid Cholesterol ester

C0322-25G C0322-5G
TCIAC0322-25GEA 84 EUR
TCIAC0322-25G TCIAC0322-5G
Cholesterol Palmitate ≥95.0% (by titrimetric analysis)
Cholesterol Palmitate
Formula: C₄₃H₇₆O₂
MW: 625.08 g/mol
Número MDL: MFCD00003638
Núm. CAS: 601-34-3
EINECS: 210-002-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Ester Value) min. 95.0 %
Specific rotation [a]20/D -22.0 to -25.0 deg (C=2, CHCl3)
Solubility in Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR