Cobalt(II) acetylacetonate dihydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Bis(2,4-pentanedionato)cobalt(II) dihydrate, Acetylacetone Cobalt(II) salt dihydrate

C0373-25G C0373-500G
TCIAC0373-25GEA 44 EUR
TCIAC0373-25G TCIAC0373-500G
Cobalt(II) acetylacetonate dihydrate ≥98.0% (by titrimetric analysis)
Cobalt(II) acetylacetonate dihydrate
Formula: C₁₀H₁₄CoO₄·2H₂O
MW: 293.18 g/mol
Número MDL: MFCD00000014
Núm. CAS: 123334-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f667b42bdf23ac2ee67b78fe8044aa0d5cedeef&vid=2ab8323c825471ebffb5dfe296003fd6792893a5&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 97.0% (calcd.on anh.substance)
Water 10.0 to 14.0 %
Solubility in hot Chloroform very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR