Picrotoxin (picrotoxinin + picrotin) ≥94.0% (by HPLC)

Proveedor: TCI
Peligro

C0375-1G C0375-5G
TCIAC0375-1GEA 62 EUR
TCIAC0375-1G TCIAC0375-5G
Picrotoxin (picrotoxinin + picrotin) ≥94.0% (by HPLC)
Picrotoxin (picrotoxinin + picrotin)
Formula: C₃₀H₃₄O₁₃
Número MDL: MFCD00074824
Núm. CAS: 124-87-8
EINECS: 204-716-6
UN: 2811
ADR: 6.1,II
Índice Merck: 14,07415

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 94.0 % (Picrotoxinin and Picrotin mixture)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR