Tubocurarine chloride pentahydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

C0433-1G C0433-100MG
TCIAC0433-1GEA 238 EUR
TCIAC0433-1G TCIAC0433-100MG
Tubocurarine chloride pentahydrate ≥98.0% (by HPLC, titration analysis)
Tubocurarine chloride pentahydrate
Formula: C₃₈H₄₄Cl₂N₂O₆
Pto. de fusión: 268°C
Número MDL: MFCD00013471
Núm. CAS: 6989-98-6
UN: 1544
ADR: 6.1,III
Índice Merck: 14,09807

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +190.0 to +195.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR