N-Cyclohexylsulphamic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

C0486-25G C0486-500G
TCIAC0486-25GEA 17 EUR
TCIAC0486-25G TCIAC0486-500G
N-Cyclohexylsulphamic acid ≥98.0% (by titrimetric analysis)
N-Cyclohexylsulphamic acid
Formula: C₆H₁₃NO₃S
Número MDL: MFCD00065234
Núm. CAS: 100-88-9
EINECS: 202-898-1
Índice Merck: 14,02703

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR