Sodium octanohydroxamate hydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Octanohydroxamic Acid Sodium Salt
C0557-1G
C0557-25G
TCIAC0557-1GEA
40
EUR
TCIAC0557-1G
TCIAC0557-25G
Sodium octanohydroxamate hydrate ≥98.0% (by titrimetric analysis)
Sodium octanohydroxamate hydrate
Formula:
C₈H₁₇NO₂ MW: 181,21 g/mol |
Número MDL:
MFCD00058968 Núm. CAS: 50292-00-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in Water | almost transparency |
Water | 7.0 to 12.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...