N-Cbz-L-asparagine ≥99.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Z-Asn-OH, Nalpha-Cbz-L-asparagine

C0573-1G C0573-10G
TCIAC0573-1GEA 16 EUR
TCIAC0573-1G TCIAC0573-10G
N-Cbz-L-asparagine ≥99.0% (by titrimetric analysis)
N-Cbz-L-asparagine
Formula: C₁₂H₁₄N₂O₅
MW: 266,3 g/mol
Número MDL: MFCD00008035
Núm. CAS: 2304-96-3
EINECS: 218-969-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Specific rotation [a]20/D +5.8 to +6.8 deg (C=1.6, AcOH)
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR