N-Carbobenzyloxy-L-alanine ≥99.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine

C0633-10G C0633-25G
TCIAC0633-10GEA 23 EUR
TCIAC0633-10G TCIAC0633-25G
N-Carbobenzyloxy-L-alanine ≥99.0% (by HPLC, titration analysis)
N-Carbobenzyloxy-L-alanine
Formula: C₁₁H₁₃NO₄
MW: 223,23 g/mol
Número MDL: MFCD00002640
Núm. CAS: 1142-20-7
EINECS: 214-532-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 99.0 %
Melting point 84.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR