N-Carbobenzoxy-D-glutamic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

C0663-25G C0663-5G
TCIAC0663-25GEA 92 EUR
TCIAC0663-25G TCIAC0663-5G
N-Carbobenzoxy-D-glutamic acid ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-D-glutamic acid
Formula: C₁₃H₁₅NO₆
MW: 281,26 g/mol
Número MDL: MFCD00063193
Núm. CAS: 63648-73-7

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 119.0 to 121.0 deg-C
Specific rotation [a]20/D +7.0 to +8.0 deg (C=8, AcOH)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR