Cholesterol propionate 95,0 GC_ASSAY_METHOD

Proveedor: TCI

Sinónimos: Propionic Acid Cholesterol Ester

C0676-10G C0676-25G
TCIAC0676-10GEA 68 EUR
TCIAC0676-10G TCIAC0676-25G
Cholesterol propionate 95,0 GC_ASSAY_METHOD
Cholesterol propionate
Formula: C₃₀H₅₀O₂
MW: 442.73 g/mol
Número MDL: MFCD00021143
Núm. CAS: 633-31-8
EINECS: 211-191-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Mesomorphic range 92.0 to 117.0 deg-C
Specific rotation [a]20/D -38.0 to -42.0 deg (C=2, CHCl3)
Solubility in Chloroform very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR