(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 98,0 HPLC, T

Proveedor: TCI

Sinónimos: Cbz-Glu-OH, Z-Glu-OH, N-Cbz-L-glutamic Acid

C0734-100G C0734-25G
TCIAC0734-100GEA 489 EUR
TCIAC0734-100G TCIAC0734-25G
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid 98,0 HPLC, T
(S)-2-(((Benzyloxy)carbonyl)amino)pentanedioic acid
Formula: C₁₃H₁₅NO₆
MW: 281.3 g/mol
Número MDL: MFCD00002801
Núm. CAS: 1155-62-0
EINECS: 214-584-1

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C
Specific rotation [a]20/D -7.0 to -8.0 deg (C=8, AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR