N-Carbobenzoxy-L-leucine ≥96.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
N-Cbz-L-leucine, Z-Leu-OH
C0739-25G
C0739-5G
TCIAC0739-25GEA
157
EUR
TCIAC0739-25G
TCIAC0739-5G
N-Carbobenzoxy-L-leucine ≥96.0% (by titrimetric analysis)
N-Carbobenzoxy-L-leucine
Formula:
C₁₄H₁₉NO₄ MW: 265,31 g/mol |
Número MDL:
MFCD00026494 Núm. CAS: 2018-66-8 EINECS: 217-960-3 |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (Neutralization titration) | min. 96.0 % |
Specific rotation [a]20/D | -15.0 to -18.0 deg (C=5, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...