4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine ≥99.0% (by titrimetric analysis) for colorimetric analysis of Co, Cd

Proveedor: TCI
Atención

Sinónimos: 4-((5-Chloropyridin-2-yl)diazenyl)benzene-1,3-diamine

C0932-1G C0932-100MG
TCIAC0932-1GEA 292 EUR
TCIAC0932-1G TCIAC0932-100MG
4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine ≥99.0% (by titrimetric analysis) for colorimetric analysis of Co, Cd
4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine
Formula: C₁₁H₁₀ClN₅
MW: 247,69 g/mol
Número MDL: MFCD00059781
Núm. CAS: 33006-91-6
EINECS: 251-334-0

Order Now

Resultados test especificaciones

Appearance Orange to Amber to Dark red powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Molar extinction coefficient (Co complex) min. 100000 (568.0 to 571.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR