Chelidonic acid monohydrate ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Oxo-4H-pyran-2,6-dicarboxylic acid monohydrate, 2,6-Dicarboxy-4-oxo-4H-pyran monohydrate

C0955-25G C0955-5G
TCIAC0955-25GEA 347 EUR
TCIAC0955-25G TCIAC0955-5G
Chelidonic acid monohydrate ≥95.0% (by titrimetric analysis)
Chelidonic acid monohydrate
Formula: C₇H₄O₆.H₂O
MW: 202,12 g/mol
Número MDL: MFCD00149433
Núm. CAS: 6003-94-7
Índice Merck: 14,02052

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 95.0% (calcd.on anh.substance)
Solubility in hot Water very faint turbidity
Water 8.5 to 9.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR