1,3,5-Cyclohexanetriol (cis and trans mixture) cis- and trans-mixture ≥95.0% (por GC)
Proveedor: TCI
C1051-25G
C1051-5G
TCIAC1051-25GEA
197
EUR
TCIAC1051-25G
TCIAC1051-5G
1,3,5-Cyclohexanetriol (cis and trans mixture) cis- and trans-mixture ≥95.0% (por GC)
1,3,5-Cyclohexanetriol (cis and trans mixture)
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol |
Número MDL:
MFCD00070580 Núm. CAS: 2041-15-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...