Comanic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Oxo-4H-pyran-2-carboxylic Acid, gamma-Pyrone-2-carboxylic Acid
C1104-1G
C1104-25G
C1104-5G
TCIAC1104-1GEA
23
EUR
TCIAC1104-1G
TCIAC1104-25G
TCIAC1104-5G
Comanic acid ≥98.0% (by GC, titration analysis)
Comanic acid
Formula:
C₆H₄O₄ MW: 140,09 g/mol |
Número MDL:
MFCD00059958 Núm. CAS: 499-05-8 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...