Direct Blue 86

Proveedor: TCI

Sinónimos: (Dihydrogenphthalocyaninedisulphonato)-coppe disodium salt

C1118-25G C1118-100G
TCIAC1118-25GEA 26 EUR
TCIAC1118-25G TCIAC1118-100G
Direct Blue 86
Direct blue 86
Formula: C₃₂H₁₆CuN₈O₆S₂
Número MDL: MFCD00070592
Núm. CAS: 1330-38-7
EINECS: 215-537-8

Order Now

Resultados test especificaciones

Appearance Dark red to Dark purple to Dark blue powder to crystal
Lambda max. 660.0 to 664.0 nm (H2O)
Lambda max.2 620.0 to 624.0 nm (H2O)
Absorbance (E1%1cm) min. 210 (H2O, 660.0 to 664.0 nm) (calcd.on dried substance)
Absorbance (E1%1cm)2 min. 250 (H2O, 620.0 to 624.0 nm) (calcd.on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR