1,2,3-Cyclohexanetriol (cis and trans mixture) cis- and trans-mixture ≥97.0% (por GC)
Proveedor: TCI
C1132-10G
C1132-25G
TCIAC1132-10GEA
118
EUR
TCIAC1132-10G
TCIAC1132-25G
1,2,3-Cyclohexanetriol (cis and trans mixture) cis- and trans-mixture ≥97.0% (por GC)
1,2,3-Cyclohexanetriol (cis and trans mixture)
Formula:
C₆H₁₂O₃ |
Número MDL:
MFCD00060111 Núm. CAS: 6286-43-7 |
Resultados test especificaciones
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 97.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...