Copper(I) trifluoromethanethiolate ≥90.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
(Trifluoromethylthio)copper(I), Copper(I) Trifluoromethanethiolate, Trifluoromethanethiol Copper(I) Salt
C1159-1G
C1159-5G
TCIAC1159-1GEA
232
EUR
TCIAC1159-1G
TCIAC1159-5G
Copper(I) trifluoromethanethiolate ≥90.0% (by titrimetric analysis)
Copper(I) trifluoromethanethiolate
Formula:
CCuF₃S MW: 164,61 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00059046 Núm. CAS: 3872-23-9 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (Chelometric Titration) | min. 90.0 % |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...