(3-Chloro-2-hydroxypropyl)trimethylammonium chloride ∼65% in water

Proveedor: TCI
Atención

Sinónimos: 3-Chloro-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride(65% in water), 3-Chloro-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride

C1180-25ML C1180-500ML
TCIAC1180-25MLEA 18 EUR
TCIAC1180-25ML TCIAC1180-500ML
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride ∼65% in water
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride
Formula: C₆H₁₅Cl₂NO
MW: 188,09 g/mol
Número MDL: MFCD00055655
Núm. CAS: 3327-22-8
EINECS: 222-048-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca7011b2981520bd8038b2384db09a3bbe40e6a5&vid=6dae42750d5660a466037c2c39e60bfe509f83e7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow clear liquid
Concentration (Argentmetric Titration) 65.0 to 73.0 w/w%
Concentration (Nonaqueous Titration) 65.0 to 73.0 w/w%
Water max. 35.0 %
pH 4.8 to 6.4 (50g/L, 25deg-C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR