Octanohydroxamic acid ≥99.0% (by total nitrogen basis)

Proveedor: TCI

C1232-25G C1232-5G
TCIAC1232-25GEA 106 EUR
TCIAC1232-25G TCIAC1232-5G
Octanohydroxamic acid ≥99.0% (by total nitrogen basis)
Octanohydroxamic acid
Formula: C₈H₁₇NO₂
Número MDL: MFCD00143918
Núm. CAS: 7377-03-9
EINECS: 230-936-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0b6a7a789e98a99adf6d354ee187b4d023d0a92&vid=f0b6a7a789e98a99adf6d354ee187b4d023d0a92&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 99.0 %
Melting point 77.0 to 82.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR