2-Methylindole-3-carboxamide ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 3-Carbamoyl-2-methylindole

C1255-1G C1255-5G
TCIAC1255-1GEA 95 EUR
TCIAC1255-1G TCIAC1255-5G
2-Methylindole-3-carboxamide ≥98.0% (by titrimetric analysis)
2-Methylindole-3-carboxamide
Formula: C₁₀H₁₀N₂O
MW: 174,20 g/mol
Número MDL: MFCD00143101
Núm. CAS: 67242-60-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 224.0 to 227.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR