(1S,2R)-(-)-10,2-Camphorsultam ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
(-)-10,2-Camphorsultam, (2R)-(-)-Bornane-10,2-sultam, (1S,5R)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.0(1,5)]decane 3,3-dioxide, (1S)-(−)-2,10-Camphorsultam, [3aS-(3aalpha,6alpha,7abeta)]-Hexahydro-8,8-dimethyl-2,2-dioxide-3H-3a,6-methano-2,1-benzisothiazole, (3aS,6R,7aR)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (3aS,6S,7aS)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (1S,2R,4R)-2,10-camphorsultam, (-)-exo-10,2-Bornanesultam, (2R)-Bornane-10,2-sultam
C1325-1G
C1325-5G
TCIAC1325-1GEA
16
EUR
TCIAC1325-1G
TCIAC1325-5G
(1S,2R)-(-)-10,2-Camphorsultam ≥98.0% (por GC)
(1S,2R)-(-)-10,2-Camphorsultam
Formula:
C₁₀H₁₇NO₂S MW: 215,32 g/mol |
Número MDL:
MFCD00066271 Núm. CAS: 94594-90-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 182.0 to 186.0 deg-C |
Specific rotation [a]20/D | -30.0 to -32.0 deg (C=1, CHCl3) |
Solubility in Chloroform | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...