Carbonylhydridotris(triphenylphosphine)rhodium(I)

Proveedor: TCI

Sinónimos: Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride, Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Hydridocarbonyltris(triphenylphosphine)rhodium(I)

C1383-1G C1383-5G
TCIAC1383-1GEA 409 EUR
TCIAC1383-1G TCIAC1383-5G
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Formula: C₅₅H₄₆OP₃Rh
MW: 918,80 g/mol
Pto. de fusión: 122°C
Número MDL: MFCD00151644
Núm. CAS: 17185-29-4
EINECS: 241-230-3
Índice Merck: 14,09758

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow powder to crystal
Elemental analysis (Carbon) 70.50 to 72.60 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR