4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole ≥97.0% (by titrimetric analysis) bifunctional fluorescent reagent

Proveedor: TCI
Peligro

Sinónimos: 4-Chloro-7-chlorosulphonylbenzofurazan, 4-Chloro-2,1,3-benzoxadiazole-7-sulphonyl Chloride

C1394-1G C1394-5G
TCIAC1394-1GEA 118 EUR
TCIAC1394-1G TCIAC1394-5G
4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole ≥97.0% (by titrimetric analysis) bifunctional fluorescent reagent
4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole
Formula: C₆H₂Cl₂N₂O₃S
MW: 253,05 g/mol
Número MDL: MFCD00191405
Núm. CAS: 142246-48-8
UN: 3261
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR