N-Carbobenzoxy-L-serine methyl ester ≥98.0% (by total nitrogen basis)
Proveedor: TCI
Sinónimos:
Z-Ser-OMe, CBZ-L-Serine methyl ester, N-Cbz-L-serine Methyl Ester, Methyl ((benzyloxy)carbonyl)-L-serinate
C1403-25G
C1403-5G
TCIAC1403-25GEA
228
EUR
TCIAC1403-25G
TCIAC1403-5G
N-Carbobenzoxy-L-serine methyl ester ≥98.0% (by total nitrogen basis)
N-Carbobenzoxy-L-serine methyl ester
Formula:
C₁₂H₁₅NO₅ MW: 253,25 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00077039 Núm. CAS: 1676-81-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 35.0 to 40.0 deg-C |
Specific rotation [a]20/D | -12.0 to -17.0 deg (C=1, MeOH) |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...