N-(Cyclohexylthio)phthalimide ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(Cyclohexylthio)isoindoline-1,3-dione, Cyclohexyl N-Phthalimidyl Sulphide
C1452-25G
C1452-500G
TCIAC1452-25GEA
18
EUR
TCIAC1452-25G
TCIAC1452-500G
N-(Cyclohexylthio)phthalimide ≥98.0% (by total nitrogen basis)
N-(Cyclohexylthio)phthalimide
Formula:
C₁₄H₁₅NO₂S MW: 261,34 g/mol |
Número MDL:
MFCD00072247 Núm. CAS: 17796-82-6 EINECS: 241-774-1 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 90.0 to 93.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...