6-Chlorohexanoic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
C1507-25G
C1507-5G
TCIAC1507-25GEA
173
EUR
TCIAC1507-25G
TCIAC1507-5G
6-Chlorohexanoic acid ≥98.0% (by GC, titration analysis)
6-Chlorohexanoic acid
Formula:
C₆H₁₁ClO₂ |
Número MDL:
MFCD00061103 Núm. CAS: 4224-62-8 UN: 3261 ADR: 8,III |
|
Resultados test especificaciones
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...