Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-(2-Aminoethyl)carbamic acid benzyl ester hydrochloride, N-1-Z-1,2-diaminoethane hydrochloride, N-Z-Ethylenediamine hydrochloride, N-Carbobenzoxy-1,2-diaminoethane hydrochloride, N-Cbz-1,2-diaminoethane hydrochloride
C1511-1G
C1511-25G
C1511-5G
TCIAC1511-1GEA
43
EUR
TCIAC1511-1G
TCIAC1511-25G
TCIAC1511-5G
Benzyl N-(2-aminoethyl)carbamate hydrochloride ≥98.0% (by titrimetric analysis)
Benzyl N-(2-aminoethyl)carbamate hydrochloride
Formula:
C₁₀H₁₄N₂O₂ MW: 230,69 g/mol Pto. de fusión: 172°C |
Número MDL:
MFCD00270150 Núm. CAS: 18807-71-1 |
Resultados test especificaciones
Appearance | White to Light yellow to Light red powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...