Sodium-4-chlorobenzenesulphinate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Sodium 4-chlorobenzenesulphinate, 4-Chlorobenzenesulphinic acid Sodium Salt
C1559-25G
C1559-5G
TCIAC1559-25GEA
81
EUR
TCIAC1559-25G
TCIAC1559-5G
Sodium-4-chlorobenzenesulphinate ≥98.0% (by titrimetric analysis)
Sodium-4-chlorobenzenesulphinate
Formula:
C₆H₅ClO₂S MW: 198,60 g/mol |
Número MDL:
MFCD00035602 Núm. CAS: 14752-66-0 EINECS: 238-811-9 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Sodium hypochlorite Method) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...