N-Carbobenzoxy-L-tyrosine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Z-Tyr-OH, N-Cbz-L-tyrosine, Cbz-Tyr-OH
C1729-25G
C1729-5G
TCIAC1729-25GEA
116
EUR
TCIAC1729-25G
TCIAC1729-5G
N-Carbobenzoxy-L-tyrosine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-L-tyrosine
Formula:
C₁₇H₁₇NO₅ MW: 315,33 g/mol |
Número MDL:
MFCD00037180 Núm. CAS: 1164-16-5 EINECS: 214-609-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Optical purity (LC) | min. 97.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...