Calcium Phthalate hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Phthalic Acid Calcium Salt

C2003-25G C2003-5G
TCIAC2003-25GEA 245 EUR
TCIAC2003-25G TCIAC2003-5G
Calcium Phthalate hydrate ≥98.0% (by titrimetric analysis)
Calcium Phthalate hydrate
Formula: C₈H₆O₄
Núm. CAS: 5793-85-1
EINECS: 227-332-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 4.0 to 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR