Cytarabine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Cytosine beta-D-Arabinofuranoside, 1-β-D-Arabinofuranosylcytosine, Arabinocytidine, Ara-C, 4-Amino-1-((2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Cytosine arabinoside, 4-Amino-1-beta-D-arabinofuranosyl-2(1H)-pyrimidinone
C2035-5G
C2035-1G
TCIAC2035-5GEA
102
EUR
TCIAC2035-5G
TCIAC2035-1G
Cytarabine ≥98.0% (by HPLC, titration analysis)
Cytarabine
Formula:
C₉H₁₃N₃O₅ MW: 243,22 g/mol Pto. de fusión: 214°C |
Número MDL:
MFCD00066487 Núm. CAS: 147-94-4 EINECS: 205-705-9 Índice Merck: 14,02784 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +155.0 to +162.0 deg (C=1, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...