Captan ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Peligro

C2059-25G C2059-5G
TCIAC2059-25GEA 149 EUR
TCIAC2059-25G TCIAC2059-5G
Captan ≥98.0% (by HPLC, total nitrogen)
Captan
Formula: C₉H₈Cl₃NO₂S
Número MDL: MFCD00041811
Núm. CAS: 133-06-2
EINECS: 205-087-0
UN: 2811
ADR: 6.1,III
Índice Merck: 14,01772

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 173.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR