N-Carbobenzoxy-D-tyrosine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (R)-2-(((Benzyloxy)carbonyl)amino)-3-(4-hydroxyphenyl)propanoic acid, N-Cbz-D-tyrosine, Z-D-Tyr-OH

C2124-1G C2124-5G
TCIAC2124-1GEA 39 EUR
TCIAC2124-1G TCIAC2124-5G
N-Carbobenzoxy-D-tyrosine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-D-tyrosine
Formula: C₁₇H₁₇NO₅
MW: 315,33 g/mol
Número MDL: MFCD00063031
Núm. CAS: 64205-12-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 96.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR