1-Cbz-piperazine ≥95.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Benzyl piperazine-1-carboxylate, Piperazine-1-carboxylic Acid Benzyl Ester, 1-piperazinecarboxylic acid, phenylmethyl ester, Benzyl 1-piperazinecarboxylate
C2178-25G
C2178-5G
TCIAC2178-25GEA
278
EUR
TCIAC2178-25G
TCIAC2178-5G
1-Cbz-piperazine ≥95.0% (by GC, titration analysis)
1-Cbz-piperazine
Formula:
C₁₂H₁₆N₂O₂ MW: 220,272 g/mol Pto. Ebullición: 161°C |
Número MDL:
MFCD00274317 Núm. CAS: 31166-44-6 |
Resultados test especificaciones
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...