Clarithromycin ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

C2220-1G C2220-25G C2220-5G
TCIAC2220-1GEA 42 EUR
TCIAC2220-1G TCIAC2220-25G TCIAC2220-5G
Clarithromycin ≥98.0% (by titrimetric analysis)
Clarithromycin
Formula: C₃₈H₆₉NO₁₃
Número MDL: MFCD00865140
Núm. CAS: 81103-11-9
Índice Merck: 14,02339

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 222.0 to 227.0 deg-C
Specific rotation [a]20/D -87.0 to -97.0 deg (C=1, CHCl3)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR