Ceftazidime (contains ca. 10% Na2CO3)

Proveedor: TCI
Peligro

C2225-25G C2225-5G
TCIAC2225-25GEA 207 EUR
TCIAC2225-25G TCIAC2225-5G
Ceftazidime (contains ca. 10% Na2CO3)
Ceftazidime
Formula: C₂₂H₂₂N₆O₇S₂
Número MDL: MFCD00072034
Núm. CAS: 72558-82-8
EINECS: 276-715-9
Índice Merck: 14,01946

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -22.0 to -26.0 deg (C=0.5, pH6.0 Phosphate Buffer)
Drying loss max. 14.0 %
Ignition residue (Sulfate) 10.0 to 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR